EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Small mammals as biomonitors of metal pollution: a case study in Slovenia.
PMID 24619365
Abstract
The transfer of lead, cadmium, zinc, mercury, copper and molybdenum from soil to the tissues of small mammals inhabiting differently polluted areas in Slovenia was investigated. Metals were determined in soil samples and in the livers of 139 individuals of five small mammal species, collected in 2012 in the vicinity of a former lead smelter, the largest Slovenian thermal power plant, along a main road and in a control area. The area in the vicinity of former lead smelter differs considerably from other study areas. The soil from that area is heavily polluted with Pb and Cd. The mean metal concentrations in the liver, irrespective of species, varied in the following ranges-Pb: 0.40-7.40 mg/kg fw and Cd: 0.27-135 mg/kg fw and reached effect concentrations at which toxic effects can be expected in a significant proportion of the livers of the small mammal specimens (Pb 40 %, Cd 67 %). These findings indicate that the majority of small mammals trapped in the area of the former lead smelter are at risk of toxic effects due to the very high bioaccumulation of Pb and Cd in the organism. On the contrary, Pd and Cd concentrations in the livers of small mammals sampled in the vicinity of the thermal power plant and along the main road were comparable with reference values and considerably lower than effect concentrations. Additionally, the study suggests that Apodemus flavicollis and Myodes glareolus are very suitable biomonitors of metal pollution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF42672427
Molybdenum, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Mo
GF11484750
Molybdenum, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Mo
GF81739457
Molybdenum, foil, not light tested, 50x50mm, thickness 0.003mm, 99.9%
Mo
GF30193781
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%
Mo
GF56008986
Molybdenum, insulated wire, 5m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF28017637
Molybdenum, microfoil, disks, 10mm, thinness 0.1μm, specific density 92.4μg/cm2, permanent mylar 3.5μm support, 99.9%
Mo
GF04272049
Molybdenum, pellets, 200g, max. size 10mm, 99.9%
Mo
GF54490574
Molybdenum, rod, 1000mm, diameter 2.0mm, centerless ground, 99.9%
Mo
GF60430766
Molybdenum, rod, 100mm, diameter 1.25mm, centerless ground, 99.9%
Mo
GF11351359
Molybdenum, wire, straight, 500mm, diameter 1.0mm, 99.95%
Mo
GF66431606
Molybdenum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Mo
GF23412383
Molybdenum, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%
Mo
GF90888788
Molybdenum, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.9%
Mo
GF98811355
Molybdenum, foil, not light tested, 25x25mm, thickness 0.01mm, annealed, 99.9%
Mo
GF26098027
Molybdenum, foil, not light tested, 50x50mm, thickness 0.001mm, 99.9%
Mo
GF63646218
Molybdenum, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.9%
Mo
GF20045786
Molybdenum, foil, not light tested, 50x50mm, thickness 0.002mm, 99.9%
Mo
GF47495975
Molybdenum, foil, not light tested, 50x50mm, thickness 0.004mm, 99.9%
Mo
GF48792628
Molybdenum, foil, not light tested, 50x50mm, thickness 0.005mm, 99.9%
Mo
GF74350267
Molybdenum, foil, not light tested, 50x50mm, thickness 0.006mm, 99.9%
Mo
GF37145358
Molybdenum, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%
Mo
GF95631419
Molybdenum, foil, not light tested, 50x50mm, thickness 0.008mm, 99.9%
Mo
GF41758768
Molybdenum, foil, not light tested, 50x50mm, thickness 0.009mm, 99.9%
Mo
GF99294968
Molybdenum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Mo
GF42252016
Molybdenum, foil, not light tested, 50x50mm, thickness 0.01mm, annealed, 99.9%
Mo
GF01123489
Molybdenum, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF40435972
Molybdenum, insulated wire, 20m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF54497111
Molybdenum, insulated wire, 50m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Mo
GF33272333
Molybdenum, mesh, 100x100mm, nominal aperture 0.44mm, wire diameter 0.07mm, 50 wires/inch, open area 67%, plain weave mesh
Mo
GF01979477
Molybdenum, mesh, 100x100mm, nominal aperture 0.8mm, wire diameter 0.18mm, 26 wires/inch, open area 67%, plain weave mesh
Mo