EMAIL THIS PAGE TO A FRIEND
American journal of clinical pathology
Mass spectrometry measurements of prostate-specific antigen (PSA) peptides derived from immune-extracted PSA provide a potential strategy for harmonizing immunoassay differences.
PMID 24619754
Abstract
Harmonization of prostate-specific antigen (PSA) immunoassays is important for good patient care. The specificity of the antibodies used to detect circulating PSA could cause differences in the PSA measurements. We used mass spectrometry (MS) to quantitate the concentration of five peptides cleaved from trypsin digestion of PSA and compared these measurements with six automated immunoassays. Linear regression and a mixed-effects model were used to analyze the results. PSA measurements from the immunoassays and the five MS peptide assays were highly correlated (R(2) > 0.99), but the recovery of the World Health Organization standard and the regression slopes differed across assays. The same relative patterns of immunoassay differences were seen in comparing their results with each of the five MS peptide measurements from different parts of the circulating PSA molecules. Mass spectrometry quantitation of peptides derived from trypsin digestion of immune-extracted PSA could be used to harmonize PSA immunoassays.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6763 Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
T8003
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
T1005
Trypsin from bovine pancreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
T8658
Trypsin from bovine pancreas, suitable for protein sequencing, lyophilized powder
T7309
Trypsin from bovine pancreas, ≥2,500 USP units/mg solid, meets USP testing specifications
T9935
Trypsin from bovine pancreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
T8802
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
T9201
Trypsin from bovine pancreas, powder, ≥7,500 BAEE units/mg solid
T6424 Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
T7409 Trypsin from porcine pancreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
T5266 Trypsin from porcine pancreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
T7168 Trypsin from porcine pancreas, tablet, 1 mg tablet
93615 Trypsin from porcine pancreas, ~1500 U/mg
85450C Trypsin Powder, Porcine 1:250, Gamma irradiated by SER-TAIN Process, dry powder
T3568 TrypZean® bovine, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)