EMAIL THIS PAGE TO A FRIEND
The journal of spinal cord medicine
Spinal cord astrocytoma mimicking multifocal myelitis.
PMID 24621037
Abstract
Differential diagnosis of acute/subacute intrinsic spinal cord lesions can be challenging. In addition, intramedullary neoplasms typically show gadolinium enhancement, mass effect, and cord expansion. We report a patient with spinal cord and brain stem lesions resembling multifocal myelitis. Magnetic resonance imaging showed no spinal cord enlargement or gadolinium enhancing. Treatment of myelitis was undertaken without stopping the progression of the disease. Biopsy was made and led to a histological diagnosis of astrocytoma. Astrocytoma must remain as a possible diagnosis of spinal cord lesions, even without typical characteristics of neoplasms. Furthermore, biopsy should always be considered when diagnosis is uncertain.