EMAIL THIS PAGE TO A FRIEND
Transplantation
Pharmacotherapy of Small-for-Size of the Liver.
PMID 24621540
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V6879
[Lys8]-Vasopressin, ≥98% (HPLC)
C46H65N13O12S2