EMAIL THIS PAGE TO A FRIEND
Inflammation
Inhibitory effects of rutin on the endothelial protein C receptor shedding in vitro and in vivo.
PMID 24622777
Abstract
Endothelial cell protein C receptor (EPCR) has important functions in regulation of coagulation and inflammation. EPCR shedding from the cell surface is mediated by tumor necrosis factor-α converting enzyme (TACE). Rutin is one of the major flavonoids from the buckwheat plant Fagopyrum tataricum. In this study, we investigated the effects of rutin on phorbol-12-myristate 13-acetate (PMA), tumor necrosis factor (TNF)-α, interleukin (IL)-1β, and on cecal ligation and puncture (CLP)-mediated EPCR shedding. We used a CLP model because this model more closely resembles human sepsis. Data showed rutin was a potent inhibitor of PMA, TNF-α, IL-1β, and CLP-induced EPCR shedding by suppression of TACE expression. Treatment with rutin resulted in a decrease of PMA-stimulated phosphorylation of p38, extracellular regulated kinases 1/2, and c-Jun N-terminal kinase. These results suggest the potential application of rutin for treatment of PMA and CLP-mediated EPCR shedding.