EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
Endogenous Tim-1 promotes severe systemic autoimmunity and renal disease MRL-Fas(lpr) mice.
PMID 24623145
Abstract
The T-cell immunoglobulin mucin 1, also known as kidney injury molecule-1, modulates CD4+ T-cell responses and is also expressed by damaged proximal tubules within the kidney. Both Th subset imbalance (Th1/Th2/Th17) and regulatory T-cell and B-cell alterations contribute to the pathogenesis of autoimmune disease. This study investigated the effects of an inhibitory anti-T-cell immunoglobulin mucin 1 antibody (RMT1-10) in lupus-prone MRL-Fas(lpr) mice. MRL-Fas(lpr) mice were treated with RMT1-10 or a control antibody intraperitoneally twice weekly from 3 mo of age for 16 wk. RMT1-10 treatment significantly improved survival, limited the development of lymphadenopathy and skin lesions, preserved renal function and decreased proteinuria, reduced serum anti-DNA antibody levels, and attenuated renal leukocyte accumulation. Th1 and Th17 cellular responses systemically and intrarenally were reduced, but regulatory T and B cells were increased. RMT1-10 treatment also reduced glomerular immunoglobulin and C3 deposition and suppressed cellular proliferation and apoptosis. Urinary excretion and renal expression of kidney injury molecule-1 was reduced, reflecting diminished interstitial injury. As RMT1-10 attenuated established lupus nephritis, manipulating immune system T-cell immunoglobulin mucin 1 may represent a therapeutic strategy in autoimmune diseases affecting the kidney.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405577
Anti-CD8A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA037756
Anti-CD8A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4700363 Monoclonal Anti-CD25-FITC antibody produced in mouse, clone MEM-181, purified immunoglobulin, buffered aqueous solution
SAB4700059
Monoclonal Anti-CD4 , (N-terminal) antibody produced in mouse, clone MEM-241, purified immunoglobulin, buffered aqueous solution
C1805 Monoclonal Anti-CD4 antibody produced in mouse, clone Q4120, purified immunoglobulin, buffered aqueous solution
SAB4700054 Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-16, purified immunoglobulin, buffered aqueous solution
SAB4700065
Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-115, purified immunoglobulin, buffered aqueous solution
SAB4700733 Monoclonal Anti-CD4 antibody produced in mouse, clone OX-35, purified immunoglobulin, buffered aqueous solution
SAB4100489
Monoclonal Anti-CD4 antibody produced in mouse, clone CD4 -A409, culture supernatant
AMAB90754
Monoclonal Anti-CD4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0395, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700060
Monoclonal Anti-CD4-APC , (N-terminal) antibody produced in mouse, clone MEM-241, purified immunoglobulin, buffered aqueous solution
SAB4700557 Monoclonal Anti-Cd4-APC antibody produced in rat, clone GK1.5, purified immunoglobulin, buffered aqueous solution
C7423
Monoclonal Anti-CD8 antibody produced in mouse, clone UCHT-4, purified immunoglobulin, buffered aqueous solution
SAB4700084 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-31, purified immunoglobulin, buffered aqueous solution
SAB4700089 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-87, purified immunoglobulin, buffered aqueous solution
SAB1402142
Monoclonal Anti-CD8A antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution