EMAIL THIS PAGE TO A FRIEND
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association
Good vs. poor self-rated diabetes control: differences in cardiovascular risk and self-care activities.
PMID 24623501
Abstract
The aim of this study was to assess differences in cardiovascular risk and performance of self-care activities in people who rated their diabetes control as good or poor. A sub-sample of 77 participants who took part in the Evaluation of Diabetes Treatment telephone interview were invited into a clinic to complete a series of laboratory examinations. Self-rated diabetes control was validated using the following laboratory markers: HbA1c, total cholesterol/HDL cholesterol ratio and LDL cholesterol. Differences in blood pressure and BMI were also assessed. Finally, all participants also completed the Summary of Self-Care activities questionnaire. Those people who rated their diabetes control as fair or poor had a significantly higher BMI, HbA1c levels, total cholesterol/HDL-cholesterol ratio and systolic blood pressure. When asked about self-care activities in the past week, those people who reported their diabetes control was fair/poor had spent significantly fewer days following a general diet and exercising. People with poor self-rated diabetes control have unfavourable cardiovascular risk and decreased performance of self-care activities.