EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
Bone- and cartilage-protective effects of a monoclonal antibody against colony-stimulating factor 1 receptor in experimental arthritis.
PMID 24623505
Abstract
Colony-stimulating factor 1 receptor (CSF-1R) essentially modulates monocyte proliferation, migration, and activation, which are considered important for the pathogenesis of rheumatoid arthritis (RA). We undertook this study to determine CSF-1R expression in human RA as well as the efficacy of a specific anti-CSF-1R monoclonal antibody (AFS98) in 2 different animal models of RA. CSF-1R expression was examined in blood, synovium, and bone samples from RA patients, osteoarthritis (OA) patients, and healthy subjects. The efficacy of AFS98 was examined by clinical assessment, histology, and bone histomorphometry in collagen-induced arthritis (CIA) and serum-transfer arthritis. CSF-1R expression was increased in the synovium of RA patients compared to OA patients and healthy controls in fibroblast-like synoviocytes, follicular dendritic cells, macrophages, and osteoclasts. Circulating RA monocytes and neutrophils but not lymphocytes were CSF-1R+. In mice, blockade of CSF-1R abrogated cartilage damage, bone erosion, and systemic bone loss, and this was associated with the depletion of osteoclasts in both models. While blockade of CSF-1R did not affect inflammation in passive serum-transfer arthritis, it significantly reduced inflammation in CIA, and this was associated with the absence of synovial macrophages and reduced splenic CD11b+Gr-1- monocytes. CSF-1R was broadly expressed in human RA. Blockade of CSF-1R protected against bone and cartilage destruction in both mouse models and also showed significant antiinflammatory effects in the CIA model. These data provide evidence for CSF-1R as a therapeutic target in RA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2501977
Anti-CSF1R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501384
Anti-CSF1R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA012323
Anti-CSF1R antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500500
Anti-CSFR antibody produced in rabbit, affinity isolated antibody
AV48225
Anti-VIM (AB1) antibody produced in rabbit, affinity isolated antibody
AV48226
Anti-VIM (AB2) antibody produced in rabbit, affinity isolated antibody
SAB1406585
Anti-VIM antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001762
Anti-VIM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
V4630 Anti-Vimentin antibody produced in goat, whole antiserum
SAB4300676
Anti-Vimentin antibody produced in rabbit, affinity isolated antibody
SAB4503081
Anti-Vimentin antibody produced in rabbit, affinity isolated antibody
C3662
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
C62H111N11O12
C1832
Cyclosporin A, from Tolypocladium inflatum, BioReagent, for molecular biology, ≥95%
C62H111N11O12
30024
Cyclosporin A, ≥98.5% (HPLC)
C62H111N11O12
32425
Cyclosporin A, VETRANAL, analytical standard
C62H111N11O12
PHR1092
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
C62H111N11O12
WH0001436M1
Monoclonal Anti-CSF1R antibody produced in mouse, clone 1G4, purified immunoglobulin, buffered aqueous solution
SAB1403703
Monoclonal Anti-CSF1R antibody produced in mouse, clone 3G12, purified immunoglobulin, buffered aqueous solution
WH0007431M1
Monoclonal Anti-VIM antibody produced in mouse, clone 3E9, purified immunoglobulin, buffered aqueous solution
SAB4700524 Monoclonal Anti-VIM antibody produced in mouse, clone VI-RE/1, purified immunoglobulin, buffered aqueous solution
SAB4100612
Monoclonal Anti-VIM antibody produced in mouse, clone VIM-A604, culture supernatant
AMAB90516
Monoclonal Anti-VIM antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0157, purified immunoglobulin, buffered aqueous glycerol solution
V6389
Monoclonal Anti-Vimentin antibody produced in mouse, ~1 mg/mL, clone V9, purified immunoglobulin, buffered aqueous solution
V5255
Monoclonal Anti-Vimentin antibody produced in mouse, clone VIM-13.2, ascites fluid
V2258
Monoclonal Anti-Vimentin antibody produced in mouse, clone LN-6, ascites fluid
PLA0199
Rabbit anti-Vimentin Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.