EMAIL THIS PAGE TO A FRIEND
Revista brasileira de hematologia e hemoterapia
Reticulocyte maturity indices in iron deficiency anemia.
PMID 24624032
Abstract
The aim of this study was to analyze the reticulocyte maturity indices (low, medium, and high fluorescence ratios) in iron deficient 1- to 6-year-old children, and identify the prevalence of iron deficiency anemia in this population. The present study included 39 subjects, divided into two groups: control subjects (n = 33), and subjects with iron deficiency anemia (n = 6). The results were analyzed by Student's t-test for comparison of means. Differences were considered significant when two-tailed p-value < 0.05. Subjects with iron deficiency anemia presented increases in the proportion of mean (10.3 ± 4.7% vs. 6.0 ± 3.4%; p-value = 0.003), and high fluorescence reticulocytes (2.3 ± 0.87% vs. 0.9 ± 0.9%; p-value = 0.03) compared to the control group. The prevalence of anemia in this population was 15% (n = 6). The indices related to immaturity of reticulocytes are higher in the presence of iron deficiency, thus demonstrating a deficiency in the raw material to form hemoglobin and are, therefore, possible early markers of iron deficiency and anemia. We emphasize the need to standardize these indices for use in clinical practice and lab test results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8173
GW833972A, ≥98% (HPLC), powder
C18H13ClF3N5O · HCl