EMAIL THIS PAGE TO A FRIEND
Radiation research
(28)silicon radiation impairs neuronal output in CA1 neurons of mouse ventral hippocampus without altering dendritic excitability.
PMID 24625098
Abstract
An unavoidable complication of space travel is exposure to radiation consisting of high-energy charged particles (HZE), such as Fe and Si nuclei. HZE radiation can affect neuronal functions at the level of the synapse or neuronal soma without inducing significant neuronal death. Different radiation species impart distinct patterns of radiation damage depending on their track structure, dose rate and fluence. Moreover, structural differences exist along the dorsoventral axis of the hippocampus that may underlie different radiosensitivities within the same neuronal field (e.g., the CA1 pyramidal cell population of the hippocampus). In this study we tested the functional effects of low doses of (28)Si radiation on excitability and synaptic plasticity in hippocampal slices prepared strictly from the ventral hippocampus. We used extracellular electrophysiological techniques to record field excitatory postsynaptic potentials (EPSPs) and population spikes in hippocampal CA1 neurons from C57BL/6J male mice 3 months after exposure to (28)Si radiation (600 MeV/n; 0.25 and 1 Gy, whole body). In irradiated mice we found prominent decrements in population spike amplitudes and reduced maximal neuronal output without changes in dendritic field EPSP. Reduced field EPSP vs. population spike ratios indicate radiation-induced impairment of the EPSP-spike (E-S) coupling. This effect was not associated with significant changes in the magnitude of short- and long-term synaptic plasticity [long-term potentiation (LTP)]. These data confirm that irradiation with (28)Si particles at relatively low doses alters the properties of the hippocampal network, which can limit its connectivity with other brain centers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF37993595
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Si
GF64686998
Silicon, rod, 100mm, diameter 2.0mm, crystalline, 100%
Si
GF22929307
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Si
GF16363391
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Si
GF66465276
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Si
GF43555664
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF70438518
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF05482852
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Si
GF91515034
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF14076930
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Si
GF28448691
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Si
GF40945754
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Si
GF64342188
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Si
GF10907377
Silicon, rod, 80mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF91141792
Silicon, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Si
GF71817543
Silicon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Si
GF75683212
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -100, 100%
Si
GF84840004
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -111, 100%
Si
GF20311254
Silicon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Si
GF41762493
Silicon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%
Si
GF85621923
Silicon, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Si
GF73529051
Silicon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Si
GF23955622
Silicon, single crystal, -100, n-type, phosphorus doped, diameter 150mm, length 0.65mm, 100%
Si
GF06693756
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.50mm, 100%
Si
GF68832805
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.75mm, 100%
Si
GF52781520
Silicon, sphere, 10pcs, diameter 2.0mm, precision sphere grade 25
Si
GF68982834
Silicon, sphere, 20pcs, diameter 2.0mm, precision sphere grade 25
Si
GF54682050
Silicon, sphere, 50pcs, diameter 2.0mm, precision sphere grade 25
Si
GF72027266
Silicon, disks, 13mm, thickness 0.38mm, single crystal, 100%
Si
GF16286227
Silicon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%
Si