EMAIL THIS PAGE TO A FRIEND
Gaoxiong yi xue ke xue za zhi = The Kaohsiung journal of medical sciences
Chemical modification of cobrotoxin with bifunctional reagent, 1,5-difluoro-2,4-dinitrobenzene.
PMID 2462645
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D102504
1,5-Difluoro-2,4-dinitrobenzene, 97%
C6H2F2N2O4