EMAIL THIS PAGE TO A FRIEND
British journal of haematology
High index of suspicion for early diagnosis of alendronate-induced stage zero osteonecrosis of jaw in thalassaemia major.
PMID 24628574
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1012780
Alendronate sodium, United States Pharmacopeia (USP) Reference Standard
C4H12NaNO7P2 · 3 H2O