EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Rapid and sensitive detection of the inhibitive activities of acetyl- and butyryl-cholinesterases inhibitors by UPLC-ESI-MS/MS.
PMID 24631841
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8375
Eserine
C15H21N3O2
45710
Eserine, ≥98.0% (N)
C15H21N3O2
51330
Harmaline, ≥95%
C13H14N2O
286044
Harmine, 98%
C13H12N2O