EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Nanostructures formed by cyclodextrin covered aminobenzophenones through supramolecular self assembly.
PMID 24632156
Abstract
Cyclodextrin (α and β) based nanostructures formed with 2-aminobenzophenone, 3-aminobenzophenone through the supramolecular self assembly are studied by absorption, fluorescence, time-resolved fluorescence, SEM, TEM, FT-IR, DSC, PXRD and (1)H NMR. The unequal layer by layer nanosheets and nanoribbons are formed through self assembly of 3ABP/CD inclusion complexes. 2ABP/α-CD complex nanostructures show the self assembly hierarchical thread structure and β-CD complexes displays a nanobrick structure. The formation of nanostructures are prearranged to HO⋯H, NH2⋯O and H2N⋯H intermolecular hydrogen bond between individual complexes. The absorption and fluorescence spectral changes explicit formation of 1:1 inclusion complexes and solvent study demonstrate the ESIPT and TICT present in both molecules. The thermodynamic parameters (ΔH, ΔG and ΔS) of 2ABP and 3ABP molecule and the inclusion complexes were determined from semiempirical PM3 calculations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
255254
3-Aminobenzophenone, 97%
C13H11NO