EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Polymer conjugates of doxorubicin bound through an amide and hydrazone bond: Impact of the carrier structure onto synergistic action in the treatment of solid tumours.
PMID 24632485
Abstract
In this study, we describe the synthesis, physico-chemical characterisation and results of the in vitro and in vivo evaluation of the biological behaviour of N-(2-hydroxypropyl)methacrylamide-based (HPMA) copolymer conjugates bearing doxorubicin (DOX) partly bound via a pH-sensitive hydrazone and partly via enzymatically degradable amide bonds, each contributing to a different anti-tumour mechanism of action of the polymer-doxorubicin conjugate. The following two types of HPMA copolymer drug carriers designed for passive tumour targeting were synthesised and compared: the linear non-degradable copolymer and the biodegradable high-molecular-weight (HMW) diblock copolymer. The HMW diblock copolymer carrier containing a degradable disulphide bond between the polymer blocks showed a rapid degradation in a buffer containing glutathione within the first few hours of incubation. In contrast to the conjugate with the amide bond-bound DOX requiring the presence of lysosomal enzymes to release DOX, the polymer-drug conjugate with the DOX bound via a hydrazone bond released DOX by pH-sensitive hydrolysis, which was significantly faster in a buffer of pH 5.0 (intracellular pH) than pH 7.4, mimicking the conditions in the bloodstream. The significant and comparable in vivo anti-tumour activity of the diblock HMW conjugate and an equimolar mixture of the conjugates differing in the DOX attachment method along with the development of cancer resistance during treatment with these conjugates demonstrated the high potential of these compounds in the development of new nanomedicines suitable for the treatment of solid tumours.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
95343
(±)-1-Amino-2-propanol, analytical standard
C3H9NO
D5767
2,2′-Dithiodipyridine, powder
C10H8N2S2
43791
2,2′-Dithiodipyridine, ≥99.0% (GC)
C10H8N2S2
143049
Aldrithiol-2, 98%
C10H8N2S2
110248
Amino-2-propanol, 93%
C3H9NO
W396508
DL-1-Amino-2-propanol, ≥90%
C3H9NO
D1515
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
C27H29NO11 · HCl
D2975000
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C27H29NO11 · HCl
1225703
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
C27H29NO11 · HCl
44583
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
C27H29NO11 · HCl
225819
Hydrazine hydrate, reagent grade, N2H4 50-60 %
H4N2 · xH2O
53847
Hydrazine hydrate solution, puriss. p.a., 24-26% in H2O (RT)
H4N2 · H2O
18412
Hydrazine hydrate solution, 78-82% (iodometric)
H4N2 · H2O
207942
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, 98%
H4N2 · H2O
64120
Methacryloyl chloride, purum, dist., ≥97.0% (GC), contains ~0.02% 2,6-di-tert-butyl-4-methylphenol as stabilizer
C4H5ClO
523216
Methacryloyl chloride, 97%, contains ~200 ppm monomethyl ether hydroquinone as stabilizer
C4H5ClO
07270
Methyl 6-aminohexanoate hydrochloride, ≥99.0% (AT)
C7H15NO2 · HCl
P2297
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
C6H3N3O9S
92822
Picrylsulfonic acid solution, 1 M in H2O
C6H3N3O9S
B91005
tert-Butyl carbazate, 98%
C5H12N2O2
901606
tert-Butyl carbazate solution, 1.0 M in dichloromethane
74564
Trifluoroacetic acid, analytical standard
C2HF3O2
233781
Triisopropylsilane, 98%
C9H22Si