EMAIL THIS PAGE TO A FRIEND
Annals of neurology
Gp120 in the pathogenesis of human immunodeficiency virus-associated pain.
PMID 24633867
Abstract
Chronic pain is a common neurological comorbidity of human immunodeficiency virus (HIV)-1 infection, but the etiological cause remains elusive. The objective of this study was to identify the HIV-1 causal factor that critically contributes to the pathogenesis of HIV-associated pain. We first compared the levels of HIV-1 proteins in postmortem tissues of the spinal cord dorsal horn (SDH) from HIV-1/acquired immunodeficiency syndrome patients who developed chronic pain (pain-positive HIV-1 patients) and HIV-1 patients who did not develop chronic pain (pain-negative HIV-1 patients). Then we used the HIV-1 protein that was specifically increased in the pain-positive patients to generate mouse models. Finally, we performed comparative analyses on the pathological changes in the models and the HIV-1 patients. We found that HIV-1 gp120 was significantly higher in pain-positive HIV-1 patients (vs pain-negative HIV-1 patients). This finding suggested that gp120 was a potential causal factor of the HIV-associated pain. To test this hypothesis, we used a mouse model generated by intrathecal injection of gp120 and compared the pathologies of the model and the pain-positive human HIV-1 patients. The results showed that the mouse model and pain-positive human HIV-1 patients developed extensive similarities in their pathological phenotypes, including pain behaviors, peripheral neuropathy, glial reactivation, synapse degeneration, and aberrant activation of pain-related signaling pathways in the SDH. Our findings suggest that gp120 may critically contribute to the pathogenesis of HIV-associated pain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA049234
Anti-AIF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501245
Anti-DLG4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405717
Anti-DLG4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401115
Anti-DLG4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA010122
Anti-DLG4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300405
Anti-GRIN1 (Ab-896) antibody produced in rabbit, affinity isolated antibody
SAB4300538
Anti-GRIN1 (Ab-897) antibody produced in rabbit, affinity isolated antibody
SAB1410533
Anti-NDOR1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA024504
Anti-NDOR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300513
Anti-SYN1 (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB1412529
ANTI-SYN1 antibody produced in mouse, clone 3E3, purified immunoglobulin, buffered aqueous solution
SAB1412557
ANTI-SYN1 antibody produced in mouse, clone 1D10, purified immunoglobulin, buffered aqueous solution
SAB1412138
ANTI-SYN1 antibody produced in mouse, clone 3F12, purified immunoglobulin, buffered aqueous solution
SAB1412425
ANTI-SYN1 antibody produced in mouse, clone 3H6, purified immunoglobulin, buffered aqueous solution
HPA000397
Anti-SYN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AB1543
Anti-Synapsin I Antibody, serum, clone, Chemicon®
S193 Anti-Synapsin I antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2501667
Anti-UCHL1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
WH0000199M1
Monoclonal Anti-AIF1 antibody produced in mouse, clone 2A2-B6, purified immunoglobulin, buffered aqueous solution
SAB5200002
Monoclonal Anti-DLG4 antibody produced in mouse, clone 6G6, 1 mg/mL, purified immunoglobulin
SAB5200003
Monoclonal Anti-DLG4 antibody produced in mouse, clone 7E3, 1 mg/mL, purified immunoglobulin
SAB4100604
Monoclonal Anti-NDOR1 antibody produced in mouse, clone NDOR1-A590, culture supernatant
SAB1409118
Monoclonal Anti-NDOR1 antibody produced in mouse, clone 5A7, purified immunoglobulin, buffered aqueous solution
SAB1405033
Monoclonal Anti-NDOR1 antibody produced in mouse, clone 3A11, ascites fluid
WH0027158M1
Monoclonal Anti-NDOR1 antibody produced in mouse, clone 3A11, purified immunoglobulin, buffered aqueous solution
P246
Monoclonal Anti-PSD95 antibody produced in mouse, clone 7E3-1B8, purified immunoglobulin, buffered aqueous solution
SAB1404432
Monoclonal Anti-SYN1 antibody produced in mouse, clone 4F6, purified immunoglobulin, buffered aqueous solution
WH0006853M7
Monoclonal Anti-SYN1 antibody produced in mouse, clone 4H1, purified immunoglobulin, buffered aqueous solution
SAB1404510
Monoclonal Anti-UCHL1 antibody produced in mouse, clone S1, purified immunoglobulin, buffered aqueous solution
WH0007345M1
Monoclonal Anti-UCHL1 antibody produced in mouse, clone 1B8-4D2, purified immunoglobulin, buffered aqueous solution