EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Liposome encapsulation of ciprofloxacin improves protection against highly virulent Francisella tularensis strain Schu S4.
PMID 24637682
Abstract
Liposome-encapsulated ciprofloxacin for inhalation (CFI) was investigated as a putative postexposure therapeutic for two strains of Francisella tularensis. The efficacies of oral ciprofloxacin and intranasally instilled CFI could not be distinguished in a mouse model of infection with the F. tularensis live vaccine strain (LVS), where a single dose of either formulation offered full protection against a lethal challenge. However, mouse studies with the more virulent Schu S4 strain of F. tularensis demonstrated that a higher level of protection against a lethal aerosol infection is provided by CFI than by oral ciprofloxacin. In addition, using this infection model, it was possible to discriminate the efficacy of intranasally instilled CFI from that of aerosolized CFI, with aerosolized CFI providing full protection after just a single dose. The improved efficacy of CFI compared to oral ciprofloxacin is likely due to the high sustained concentrations of ciprofloxacin in the lung. In summary, CFI may be a promising therapy, perhaps enabling the prophylactic regimen to be shortened, for use in the event of a deliberate release of F. tularensis. The prophylactic efficacy of CFI against other biological warfare (BW) threat agents also warrants investigation.