EMAIL THIS PAGE TO A FRIEND
Journal of Biomedical Materials Research Part A
Optimization of release pattern of FGF-2 and BMP-2 for osteogenic differentiation of low-population density hMSCs.
PMID 24639043