EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Hydrodynamics selects the pathway for displacive transformations in DNA-linked colloidal crystallites.
PMID 24639545
Abstract
The degree to which DNA-linked particle crystals, particularly those composed of micrometer-scale colloids, are able to dynamically evolve or whether they are kinetically arrested after formation remains poorly understood. Here, we study a recently observed displacive transformation in colloidal binary superlattice crystals, whereby a body-centered cubic to face-centered cubic transformation is found to proceed spontaneously under some annealing conditions. Using a comprehensive suite of computer simulation tools, we develop a framework for analyzing the many displacive transformation pathways corresponding to distinct, but energetically degenerate, random hexagonal close-packed end states. Due to the short-ranged, spherically symmetric nature of the particle interactions the pathways are all barrierless, suggesting that all end states should be equally likely. Instead, we find that hydrodynamic correlations between particles result in anisotropic mobility along the various possible displacive pathways, strongly selecting for pathways that lead to the fcc-CuAu-I configuration, explaining recent experimental observations. This finding may provide clues for discovering new approaches for controlling structure in this emerging class of materials.