EMAIL THIS PAGE TO A FRIEND
Journal of receptor and signal transduction research
Creatinine metabolite, HMH (5-hydroxy-1-methylhydantoin; NZ-419), modulates bradykinin-induced changes in vascular smooth muscle cells.
PMID 24641698
Abstract
A creatinine metabolite, 5-hydroxy-1-methylhydantoin (HMH: NZ-419), a hydroxyl radical scavenger, has previously been shown to confer renoprotection by inhibiting the progression of chronic kidney disease in rats. In the current study, we demonstrate that HMH modulates the effects of glucose and bradykinin (BK) in vascular smooth muscle cell (VSMC). HMH a novel anti-oxidant drug completely suppressed the expression of B2-kinin receptors (B2KR) in response to high glucose (25 mM) stimulation in VSMC and was also shown to attenuate the effects of BK on VSMC remodeling. HMH inhibited the BK-induced increase in MAPK phosphorylation and attenuated the increase in connective tissue growth factor (CTGF) protein levels in VSMC. These findings suggest that HMH may confer vascular protection against high glucose concentrations and BK-stimulation to ameliorate vascular injury and remodeling through its anti-oxidant properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA050841
Anti-BDKRB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41781
Anti-BDKRB2 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB4200176
Anti-JNK antibody, Mouse monoclonal, clone 1C2, purified from hybridoma cell culture
SAB4502398
Anti-JNK1 antibody produced in rabbit, affinity isolated antibody
M7556 Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
SAB4300499
Anti-MAPK14 (Ab-182) antibody produced in rabbit, affinity isolated antibody
SAB1405665
Anti-MAPK14 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
AV06004
Anti-MAPK14 antibody produced in rabbit, affinity isolated antibody
SAB1406300
Anti-MAPK8 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4500493
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500491
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
SAB4500492
Anti-p38 MAPK antibody produced in rabbit, affinity isolated antibody
HPA025018
Anti-PLEKHM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB3500458
Anti-PLEKHM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4200544
Anti-Synaptophysin antibody, Mouse monoclonal, clone SVP-38, purified from hybridoma cell culture
HPA002858
Anti-SYP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102352
Anti-SYP antibody produced in rabbit, affinity isolated antibody
WH0000624M1
Monoclonal Anti-BDKRB2 antibody produced in mouse, clone 3F6, purified immunoglobulin, buffered aqueous solution
M7431 Monoclonal Anti-MAP Kinase 2 (ERK-2) antibody produced in mouse, clone 1B3B9, purified immunoglobulin, buffered aqueous solution
SAB1404249
Monoclonal Anti-MAPK8 antibody produced in mouse, clone 2F11, purified immunoglobulin, buffered aqueous solution
WH0009842M1
Monoclonal Anti-PLEKHM1 antibody produced in mouse, clone 1C9, purified immunoglobulin, buffered aqueous solution
WH0006855M1
Monoclonal Anti-SYP antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
WH0005599M2
Monoclonal Anti-MAPK8 antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution