EMAIL THIS PAGE TO A FRIEND
Fitoterapia
Diterpenes inhibiting NO production from Euphorbia helioscopia.
PMID 24641945
Abstract
Three new jatrophane diterpenes (1-3), an unreported spectroscopic data jatrophane diterpenene (4), and nine known analogues (5-13) have been isolated from the whole plants of Euphorbia helioscopia. Their structures were established by detailed spectroscopic data analyses (IR, ESIMS, HR-ESIMS, and 1D and 2D NMR), and the structure of 1 was confirmed by X-ray crystallography. The diterpenes showed inhibitory activities on LPS-induced NO production in murine microglial BV-2 cells.