EMAIL THIS PAGE TO A FRIEND
CNS spectrums
Efficacy of adjunctive aripiprazole in patients with major depressive disorder whose symptoms worsened with antidepressant monotherapy.
PMID 24642260
Abstract
Efficacy of depression treatments, including adjunctive antipsychotic treatment, has not been explored for patients with worsening symptoms after antidepressant therapy (ADT). This post-hoc analysis utilized pooled data from 3 similarly designed, randomized, double-blind, placebo-controlled trials that assessed the efficacy, safety, and tolerability of adjunctive aripiprazole in patients with major depressive disorder with inadequate response to ADT. The studies had 2 phases: an 8-week prospective ADT phase and 6-week adjunctive (aripiprazole or placebo) treatment phase. This analysis focused on patients whose symptoms worsened during the prospective 8-week ADT phase (worsening defined as >0% increase in Montgomery-Åsberg Depressive Rating Scale [MADRS] Total score). During the 6-week, double-blind, adjunctive phase, response was defined as ≥50% reduction in MADRS Total score and remission as ≥50% reduction in MADRS Total score and MADRS score ≤10. Of 1065 patients who failed to achieve a response during the prospective phase, 160 exhibited worsening of symptoms (ADT-Worseners), and 905 exhibited no change/reduction in MADRS scores (ADT-Non-worseners). Response rates for ADT-Worseners at endpoint were 36.6% (adjunctive aripiprazole) and 22.5% (placebo). Similarly, response rates at endpoint for ADT-Non-worseners were 37.5% (adjunctive aripiprazole) and 22.5% (placebo). Remission rates at endpoint for ADT-Worseners were 25.4% (adjunctive aripiprazole) and 12.4% (placebo). For ADT-Non-worseners, remission rates were 29.9% (adjunctive aripiprazole) and 17.4% (placebo). These results suggest that adjunctive aripiprazole is an effective intervention for patients whose symptoms worsen during antidepressant monotherapy. The results challenge the view that benefits of adjunctive therapy with aripiprazole are limited to partial responders to ADT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001649 Aripiprazole, European Pharmacopoeia (EP) Reference Standard
C23H27Cl2N3O2