EMAIL THIS PAGE TO A FRIEND
Annals of Neurology
Homocysteine exacerbates ?-amyloid pathology, tau pathology, and cognitive deficit in a mouse model of Alzheimer disease with plaques and tangles.
PMID 24644038