EMAIL THIS PAGE TO A FRIEND
The American journal of the medical sciences
Vancomycin-induced interstitial nephritis superimposed on coexisting renal disease: the importance of renal biopsy.
PMID 24646459
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V0045000
Vancomycin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C66H75Cl2N9O24 · xHCl