EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
A systemically-available kynurenine aminotransferase II (KAT II) inhibitor restores nicotine-evoked glutamatergic activity in the cortex of rats.
PMID 24647121