EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A chiral thioxanthone as an organocatalyst for enantioselective [2+2] photocycloaddition reactions induced by visible light.
PMID 24648167
Abstract
Thioxanthone 1, which was synthesized in a concise fashion from methyl thiosalicylate, exhibits a significant absorption in the visible light region. It allows for an efficient enantioselective catalysis of intramolecular [2+2] photocycloaddition reactions presumably by triplet energy transfer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357758
Methyl thiosalicylate, 97%
C8H8O2S