EMAIL THIS PAGE TO A FRIEND
Journal of environmental sciences (China)
Isolation of a salt tolerant laccase secreting strain of Trichoderma sp. NFCCI-2745 and optimization of culture conditions and assessing its effectiveness in treating saline phenolic effluents.
PMID 24649671
Abstract
Most of the hazardous pollutants are phenolic in nature and persists in the environment. The ability of laccases to oxidize phenolic compounds and reduce molecular oxygen to water has led to intensive studies of these enzymes. Therefore the fungal strains with high laccase activity and substrate affinity that can tolerate harsh environmental conditions have a potential for biotechnological applications. Salt tolerant laccase secreting fungi can be utilized in treatment of saline and phenolic rich industrial effluents such as coir effluent and textile effluent that needed to be diluted several fold before microbial treatment. This is the first study describing the isolation and optimization of a salt tolerant strain of Trichoderma sp. potential for industrial applications. The fungus was identified based on morphological characteristics and was subsequently confirmed with molecular techniques and deposited at National Fungal Culture Collections of India (NFCCI) under the Accession No. Trichoderma viride NFCCI 2745. In contrast to other laccase secreting fungi, light conditions did not exert much influence on laccase production of this strain and salinity enhanced its laccase secretion. The fungus effectively removed the phenolic content of the textile effluent, coir-ret liquor and wood processing effluent within 96 hr of incubation. The tolerance of the fungus to high salinity and phenolic compounds makes this strain ideal for treating saline and phenolic rich industrial effluents.