EMAIL THIS PAGE TO A FRIEND
Pharmaceutical biology
Isolation, characterization and antihyperlipidemic activity of secoisolariciresinol diglucoside in poloxamer-407-induced experimental hyperlipidemia.
PMID 24649910