EMAIL THIS PAGE TO A FRIEND
In vitro cellular & developmental biology. Animal
Kidney injury molecule-1 is involved in the chemotactic migration of mesenchymal stem cells.
PMID 24652046
Abstract
A better understanding of the organ specific factors that regulate the migration of mesenchymal stem cells (MSCs) into the target organ is essential for optimization of strategies to improve the repair after injury. In the present study, we showed that the kidney injury molecule-1 (KIM-1), a well-known kidney-specific biomarker, enhanced the in vitro migration capacity of MSCs as a potent kidney-specific chemo-attractant or an inducer. The in vitro roles were verified by migration assay using KIM1-PK1 cell lines, the mouse proximal tubular epithelial cells (mPTEs) and recombinant human KIM-1 proteins (rhKIM-1). Immunofluorescence staining displayed specific ectodomain binding of KIM-1 on the surface of MSCs. Upregulation of chemokine receptor type 4 (CXCR4) protein when treated with tumor necrosis factor alpha (TNF-α) was shown. The effect of KIM-1 on migration of MSCs was augmented by TNF-α pretreatment in a dose-dependent manner, and reduced by AMD3100, an antagonist of CXCR4. These results suggest that KIM-1 is a potential chemo-ligand of CXCR4 and may play an important role in kidney-specific migration of MSCs via interaction between KIM-1 and CXCR4.