EMAIL THIS PAGE TO A FRIEND
Journal of microencapsulation
Evaluation of anticancer activity of celastrol liposomes in prostate cancer cells.
PMID 24654943
Abstract
Celastrol, a natural compound derived from the herb Tripterygium wilfordii, is known to have anticancer activity, but is not soluble in water. Formation of celastrol liposomes, to avoid the use of toxic solubilising agents. Two different formulations of PEGylated celastrol liposomes were fabricated. Liposomal characteristics and serum stability were determined using dynamic light scattering. Drug entrapment efficacy and drug release were measured spectrophotometrically. Cellular internalisation and anticancer activity was measured in prostate cancer cells. Liposomal celastrol displayed efficient serum stability, cellular internalisation and anticancer activity, comparable to that of the free drug reconstituted in dimethyl sulfoxide. Liposomal celastrol can decrease the viability of prostate cancer cells, while eliminating the need for toxic solubilising agents.