EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and haemostasis : JTH
Systemic inhibition and liver-specific over-expression of PAI-1 failed to improve survival in all-inclusive populations or homogenous cohorts of CLP mice.
PMID 24655755
Abstract
The role of plasminogen activator inhibitor type-1 (PAI-1) in abdominal sepsis remains elusive. To study the influence of inhibition and over-expression of PAI-1 upon survival in cecal ligation and puncture (CLP) sepsis. (i) Mice underwent moderate CLP and received 10 mg kg(-1) of either monoclonal anti-PAI-1 (MA-MP6H6) or control (MA-Control) antibody intravenously at 0, 18 or 30 h post-CLP. The 30-h treatment group was additionally stratified into mice predicted to survive (P-SUR) or die (P-DIE) based on IL 6 measured at 24 h post-CLP. (ii) PAI-1 expression was induced with pLIVE.PAI-1 plasmid administered 72 h pre-CLP. Blood was sampled for 5 days and survival was monitored for 28 days. MA-MP6H6 effectively neutralized active PAI-1 and fully restored fibrinolysis while PAI-1 over-expression was liver-specific and correlated with PAI-1 increase in the blood. Without stratification, MA-MP6H6 co-/post-treatment conferred no survival benefit. Prospective stratification (IL-6 cut-off: 14 ng mL(-1) ) suggested increased mortality by MA-MP6H6 treatment in P-SUR that reached 30% difference (vs. MA-Control; P < 0.05) after a retrospective cut-off readjustment to 3.3 ng mL(-1) for better P-SUR homogeneity. Subsequent prospective anti-PAI-1 treatment in P-SUR mice with 3.3 ng mL(-1) cut-off demonstrated a negative but statistically insignificant effect: mortality was higher by 17% after MA-MP6H6 vs. MA-Control. Over-expression of PAI 1 did not alter post-CLP survival. Neither PAI-1 inhibition nor over-expression meaningfully modified inflammatory response and/or organ function. Restoration of fibrinolysis in early abdominal sepsis was not beneficial and it may prove detrimental in subjects with the lowest risk of death, while preemptive PAI-1 up-regulation at the current magnitude was not protective.