EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Nutrition: the future of melanoma prevention?
PMID 24656410
Abstract
Melanoma is one of the deadliest forms of skin cancer, having a high metastatic potential and afflicting all age groups. The need for successful preventative measures is particularly urgent as metastatic melanoma is largely incurable. The beneficial role of nutrition and other natural compounds in the prevention and treatment of melanoma has been clearly demonstrated in the past, and is an exciting source for potential therapies in the future. We sought to review updates in the current literature regarding new developments in the relationship between nutrition and melanoma risk and treatment. Articles in the public domain regarding the impact of diet, grape seed proanthocyanidins, selenium, vitamin D, vitamin E, epigallocatechin-3-gallate, resveratrol, rosmarinic acid, lycopene, and fig latex on melanoma were included. Grape seed proanthocyanidins, epigallocatechin-3-gallate, resveratrol, rosmarinic acid, lycopene, and fig latex have demonstrated clear anticancer effects toward melanoma. The roles of selenium, vitamin D, and vitamin E, however, have been more controversial. None. The role of natural compounds in the future of melanoma prevention and treatment is promising and one that is worthy of further exploration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
75051
Lycopene, analytical standard
C40H56