EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
A quick, easy, cheap, effective, rugged and safe extraction method followed by liquid chromatography-(Orbitrap) high resolution mass spectrometry to determine benzotriazole, benzothiazole and benzenesulfonamide derivates in sewage sludge.
PMID 24656545
Abstract
A Quick, Easy, Cheap, Effective, Rugged and Safe (QuEChERS) extraction method followed by liquid chromatography-(Orbitrap) high resolution mass spectrometry was developed for the simultaneous determination of five benzotriazole, four benzothiazole and five benzenesulfonamide derivates in sewage sludge. While the method was being developed, several buffers and dispersive solid-phase extraction clean-up (dSPE) sorbents were tested. Citrate buffer and Z-sep+ (zirconium-based sorbent) were the most effective extraction buffer and dSPE clean-up material. The absolute recoveries were higher than 80% for all compounds (100ng/g (d.w.)) and the matrix effect was less than -20% for most compounds. The limits of detection were between 0.5 and 10ng/g (d.w.) and the limits of quantification (LOQ) were between 1 and 25ng/g (d.w.). Repeatability and reproducibility were lower than 15% (%RSD, n=5). Several sludge samples from five sewage treatment plants in Catalonia were analysed and the most abundant compounds were 2-hydroxybenzothiazole (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55299-U
Supel QuE, Z-Sep+ Bulk, pkg of 20 g
55284-U
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100
55401-U
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 12 mL, pack of 50 ea
55403-U
Supel QuE, Z-Sep Tube, centrifuge tube volume 12 mL, pack of 50 ea
55406-U
Supel QuE, Z-Sep+ with MgSO4 Tube, centrifuge tube volume 12 mL, pack of 50 ea
55407-U Supel QuE, Z-Sep with MgSO4 Tube, centrifuge tube volume 12 mL, pack of 50 ea
55408-U
Supel QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
55411-U
Supel QuE, Z-Sep Tube, centrifuge tube volume 2 mL, pack of 100 ea
55414-U
Supel QuE, Z-Sep+ with MgSO4 Tube, centrifuge tube volume 2 mL, pack of 100 ea
55417-U
Supel QuE, Z-Sep with MgSO4 Tube, centrifuge tube volume 2 mL, pack of 100 ea
55418-U
Supel QuE, Z-Sep Bulk
55486-U
Supel QuE, Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
55491-U
Supel QuE, Z-Sep Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
55503-U
Supel QuE, Z-Sep with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
55506-U
Supel QuE, Z-Sep/C18 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
55511-U
Supel QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible