EMAIL THIS PAGE TO A FRIEND
Journal of assisted reproduction and genetics
Characterization of secreted proteins of 2-cell mouse embryos cultured in vitro to the blastocyst stage with and without protein supplementation.
PMID 24658922
Abstract
To identify the secreted proteins of murine embryos grown in vitro. Two-cell mouse embryos (n=432) were randomly allocated to culture to the blastocyst stage in protein-free and in protein-supplemented (3 % BSA) media. Proteins were separated by SDS-PAGE; bands were visualized by coomassie staining, followed by in-gel trypsin digestion and liquid chromatography-tandem mass spectrometry. RT-PCR and confocal microscopy were used to confirm gene/protein expression in blastocysts. Of all individually identified proteins, 34 and 23 were found in embryos cultured without and with BSA, respectively, and 20 were common. Identified proteins having an N-terminal secretory sequence or transmembrane domains located on the extracellular backbone were postulated as secreted proteins. Gene and protein expression for two selected molecules were confirmed. Functional analysis revealed over-represented processes related to lipid metabolism, cyclase activity, and cell adhesion/membrane functions. This study provided evidence to further characterize secreted proteins by mouse embryos grown from the 2-cell to the blastocyst stage in vitro. Because of homology between murine and human, these results may provide information to be translated to the clinical setting.