EMAIL THIS PAGE TO A FRIEND
Organic letters
Copper-mediated trifluoromethylthiolation of α-diazoesters.
PMID 24660960
Abstract
A novel Cu-mediated trifluoromethylthiolation of diazo compounds has been developed that provides a convenient synthetic route for the efficient α-trifluoromethylthiolation of simple esters under mild reaction conditions. The reaction is typically carried out at room temperature, and water could be used to promote the reaction.