EMAIL THIS PAGE TO A FRIEND
Journal of Ethnopharmacology
An integrated strategy based on UPLC-DAD-QTOF-MS for metabolism and pharmacokinetic studies of herbal medicines: Tibetan Snow Lotus herb (Saussurea laniceps), a case study.
PMID 24661968