EMAIL THIS PAGE TO A FRIEND
Testing
A recombinant novirhabdovirus presenting at the surface the E Glycoprotein from West Nile Virus (WNV) is immunogenic and provides partial protection against lethal WNV challenge in BALB/c mice.
PMID 24663075