EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Directed axonal outgrowth using a propagating gradient of IGF-1.
PMID 24664530
Abstract
The temporospatial regulation of axon outgrowth is useful for guiding de novo connectivity or re-connectivity of neurons in neurological injury or disease. Here we report the successful construction of a biocompatible guidance device, in which a linear propagation of IGF-1 gradient sequentially directs axon outgrowth. We observe the extensive in vitro axonal extension over 5 mm with a desired growth rate of ∼ 1 mm/day.