EMAIL THIS PAGE TO A FRIEND
Pediatric blood & cancer
Initial testing (stage 1) of the notch inhibitor PF-03084014, by the pediatric preclinical testing program.
PMID 24664981
Abstract
PF-03084014, a γ-secretase inhibitor, was tested against the PPTP in vitro cell line panel (1.0 nM to 10 μM) and against the in vivo xenograft panels (administered orally twice daily on Days 1-7 and 15-21). PF-03084014 demonstrated limited in vitro activity, with no cell line achieving ≥50% inhibition. PF-03084014 induced significant differences in EFS distribution in 14 of 35 (40%) solid tumor xenografts, and 1 of 9 ALL xenografts (which lacked a NOTCH1 mutation), but objective responses were not observed. PF-03084014 demonstrated limited single agent activity in vitro and in vivo against the pediatric preclinical models studied.