EMAIL THIS PAGE TO A FRIEND
Journal of nuclear medicine : official publication, Society of Nuclear Medicine
Glypican-3-targeted 89Zr PET imaging of hepatocellular carcinoma: where antibody imaging dares to tread.
PMID 24665087
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF69451487
Zirconium, foil, 300x300mm, thickness 0.25mm, annealed, 99.2%
Zr
GF27504402
Zirconium, foil, 300x300mm, thickness 0.5mm, annealed, 99.2%
Zr
GF74069837
Zirconium, foil, 50x50mm, thickness 0.125mm, annealed, 99.2%
Zr
GF87053474
Zirconium, foil, 50x50mm, thickness 0.25mm, annealed, 99.2%
Zr
GF83603909
Zirconium, foil, 50x50mm, thickness 0.5mm, annealed, 99.2%
Zr
GF04051886
Zirconium, foil, 50x50mm, thickness 2.0mm, annealed, 99.2%
Zr
GF14815593
Zirconium, foil, 10mm disks, thickness 0.0125mm, 99.2%
Zr
GF58113019
Zirconium, foil, 10mm disks, thickness 0.015mm, 99.2%
Zr
GF67870468
Zirconium, foil, 10mm disks, thickness 0.016mm, 99.2%
Zr
GF58442070
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF54823642
Zirconium, foil, 10mm disks, thickness 0.01mm, 99.2%
Zr
GF75562869
Zirconium, foil, 10mm disks, thickness 0.025mm, annealed, 99.2%
Zr
GF55287221
Zirconium, foil, 10mm disks, thickness 0.02mm, 99.2%
Zr
GF38009044
Zirconium, foil, 10mm disks, thickness 0.05mm, annealed, 99.2%
Zr
GF89139594
Zirconium, foil, 10mm disks, thickness 0.075mm, annealed, 99.2%
Zr
GF22007595
Zirconium, foil, 10mm disks, thickness 0.125mm, annealed, 99.2%
Zr
GF50038039
Zirconium, foil, 10mm disks, thickness 0.25mm, annealed, 99.2%
Zr
GF03886443
Zirconium, foil, 10mm disks, thickness 0.5mm, annealed, 99.2%
Zr
GF09475027
Zirconium, foil, 15mm disks, thickness 0.009mm, 99.2%
Zr
GF51948872
Zirconium, foil, 15mm disks, thickness 0.0125mm, 99.2%
Zr
GF83808221
Zirconium, foil, 15mm disks, thickness 0.016mm, 99.2%
Zr
GF05259135
Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%
Zr
GF72812370
Zirconium, foil, 15mm disks, thickness 0.01mm, 99.2%
Zr
GF45357688
Zirconium, foil, 15mm disks, thickness 0.025mm, annealed, 99.2%
Zr
GF51130370
Zirconium, foil, 15mm disks, thickness 0.05mm, annealed, 99.2%
Zr
GF42449249
Zirconium, foil, 15mm disks, thickness 0.075mm, annealed, 99.2%
Zr
GF50949232
Zirconium, foil, 15mm disks, thickness 0.125mm, annealed, 99.2%
Zr
GF24728890
Zirconium, foil, 15mm disks, thickness 0.1mm, annealed, 99.2%
Zr
GF54754218
Zirconium, foil, 15mm disks, thickness 0.25mm, annealed, 99.2%
Zr
GF36480506
Zirconium, foil, 1m coil, thickness 0.016mm, 99.2%
Zr