EMAIL THIS PAGE TO A FRIEND
Journal of Cancer
Relationship between Serum Level of Lymphatic Vessel Endothelial Hyaluronan Receptor-1 and Prognosis in Patients with Lung Cancer.
PMID 24665348
Abstract
Lymphatic vessel endothelial hyaluronan receptor-1 (LYVE-1) is a hyaluronic acid receptor that is selectively expressed in the endothelia of lymphatic capillaries. The density of lymphatic vessels expressing LYVE-1 on immunohistochemistry negatively correlates with prognosis of patients with non-small-cell lung cancer. However, the relationship between LYVE-1 serum levels and lung cancer staging is unknown. We collected blood samples from 58 lung cancer patients before treatment and measured LYVE-1 serum levels using an enzyme-linked immunosorbent assay. Mean serum LYVE-1 levels were 1,420 pg/mL. Serum LYVE-1 levels correlated positively with serum albumin levels, but inversely with primary tumor size, leukocyte counts, and platelet counts by Pearson's product-moment correlation coefficient. A high cancer staging, occurrence of lymph-node metastases, and occurrence of distant metastases were significantly associated with low LYVE-1 levels. Moreover, multiple logistic regression analyses revealed that LYVE-1 levels were predictive of the presence of lymph node and distant metastases, independently of the other factors. Kaplan-Meier analysis showed that the survival of patients with serum LYVE-1 ≤1,553 pg/mL was significantly poorer than that of patients with serum LYVE-1 >1,553 pg/mL. This survival difference relative to LYVE-1 levels remained statistically significant after adjusting for age and gender by the Cox proportional-hazard analysis. Serum LYVE-1 is significantly low in lung cancer patients with metastasis, compared with those without. Measuring LYVE-1 levels in lung cancer patients may be useful for evaluating lung cancer progression.