EMAIL THIS PAGE TO A FRIEND
The world journal of biological psychiatry : the official journal of the World Federation of Societies of Biological Psychiatry
Achievement of take-home dose privileges is associated with better-perceived sleep and with cognitive status among methadone maintenance treatment patients.
PMID 24666249
Abstract
Methadone maintenance treatment (MMT) patients may achieve up to a 2-week privilege of methadone take-home doses (THD), which is associated with considerable responsibility. MMT patients are characterized as having poor sleep quality and low cognitive states. We studied sleep indices and cognitive status with respect to THD privileges. A sample of 123 MMT patients stratified by THD groups was studied. Pittsburgh Sleep Quality Index (PSQI), Epworth Sleepiness Scale (ESS), and the cognitive Clock Drawing Test (CDT) were performed. Thirty-one of 123 patients never had any THD and 92 did (25 had the maximum of 2 weeks). The never THD had history of longer duration of opiate usage and a shorter period in MMT. They had the highest rates of poor sleep (80.6%, PSQI > 5), daily sleepiness ("fall asleep while talking") (41.9%), and impaired cognitive status (58.1%, CDT < 3), while those who had 2-week privileges had the lowest (56, 8, and 28%, respectively). Logistic regression characterized THD patients as no-benzodiazepine and no-cocaine, short opiate usage duration, low ADHD scores, and no cognitive impairment (CDT = 3) and its interaction with treatment duration. Privileges that reflect patients' abstinence and rehabilitation were also expanded to be associated with better cognitive states. These finding confirm the THD dispensing performance. Including CDT as part of the decision for dispensing THD may be considered.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
317632
(Methylcyclopentadienyl)manganese(I) tricarbonyl
C9H7MnO3