EMAIL THIS PAGE TO A FRIEND
Clinical Microbiology and Infection : the Official Publication of the European Society of Clinical Microbiology and Infectious Diseases
Epstein-Barr virus BZLF1 gene polymorphisms: malignancy related or geographically distributed variants?
PMID 24666405