EMAIL THIS PAGE TO A FRIEND
Journal of Veterinary Pharmacology and Therapeutics
Intravenous infusion of electrolyte solution changes pharmacokinetics of drugs: pharmacokinetics of ampicillin.
PMID 24666465