EMAIL THIS PAGE TO A FRIEND
Journal of surgical oncology
Clinical significance of primary tumor resection in colorectal cancer patients with synchronous unresectable metastasis.
PMID 24668581
Abstract
The aim of this study was to compare the outcomes in colorectal cancer (CRC) patients with synchronous unresectable metastases who received either primary tumor resection (PTR) or chemotherapy as the first treatment and to investigate the clinical course of asymptomatic patients who received chemotherapy as the first treatment. We retrospectively analyzed 324 CRC patients with synchronous unresectable metastases. Overall survival (OS) was analyzed for the two groups (upfront PTR group [n = 72] vs. upfront chemotherapy group [n = 252]). Surgical morbidity and mortality were recorded. In the asymptomatic patients who received upfront chemotherapy, the incidences of primary tumor-related complications were analyzed. In patients who underwent PTR as the first treatment, the median OS period was superior to those who received upfront chemotherapy (17.2 vs. 13.6 months, P = 0.002). In the PTR group, surgical morbidity and mortality were 11.6% and 1.9%, respectively. Of the 252 asymptomatic patients, the incidence of primary tumor-related complications was 35%. Emergent surgery was ultimately done in 14% of the 252 patients. CRC patients with synchronous unresectable metastases who underwent PTR followed by chemotherapy had significantly longer survival times compared to patients who received chemotherapy as the first treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0250000
Calcium folinate, European Pharmacopoeia (EP) Reference Standard
C20H21CaN7O7 · xH2O