EMAIL THIS PAGE TO A FRIEND
Oxidative Medicine and Cellular Longevity
Propofol activation of the Nrf2 pathway is associated with amelioration of acute lung injury in a rat liver transplantation model.
PMID 24669282