EMAIL THIS PAGE TO A FRIEND
Journal of neurophysiology
Synaptic excitation is regulated by the postsynaptic dSK channel at the Drosophila larval NMJ.
PMID 24671529
Abstract
In the mammalian central nervous system, the postsynaptic small-conductance Ca(2+)-dependent K(+) (SK) channel has been shown to reduce postsynaptic depolarization and limit Ca(2+) influx through N-methyl-d-aspartate receptors. To examine further the role of the postsynaptic SK channel in synaptic transmission, we studied its action at the Drosophila larval neuromuscular junction (NMJ). Repetitive synaptic stimulation produced an increase in postsynaptic membrane conductance leading to depression of excitatory postsynaptic potential amplitude and hyperpolarization of the resting membrane potential (RMP). This reduction in synaptic excitation was due to the postsynaptic Drosophila SK (dSK) channel; synaptic depression, increased membrane conductance and RMP hyperpolarization were reduced in dSK mutants or after expressing a Ca(2+) buffer in the muscle. Ca(2+) entering at the postsynaptic membrane was sufficient to activate dSK channels based upon studies in which the muscle membrane was voltage clamped to prevent opening voltage-dependent Ca(2+) channels. Increasing external Ca(2+) produced an increase in resting membrane conductance and RMP that was not seen in dSK mutants or after adding the glutamate-receptor blocker philanthotoxin. Thus it appeared that dSK channels were also activated by spontaneous transmitter release and played a role in setting membrane conductance and RMP. In mammals, dephosphorylation by protein phosphatase 2A (PP2A) increased the Ca(2+) sensitivity of the SK channel; PP2A appeared to increase the sensitivity of the dSK channel since PP2A inhibitors reduced activation of the dSK channel by evoked synaptic activity or increased external Ca(2+). It is proposed that spontaneous and evoked transmitter release activate the postsynaptic dSK channel to limit synaptic excitation and stabilize synapses.