EMAIL THIS PAGE TO A FRIEND
Contact Dermatitis
Characterization of skin sensitizers from autoxidized citronellol - impact of the terpene structure on the autoxidation process.
PMID 24673435
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27483
(−)-β-Citronellol, analytical standard
C10H20O
CDS000400
1,2,3,5-Tetramethylbenzene, AldrichCPR
C10H14
90872
Acetone, analytical standard
C3H6O
1006801
Acetone, United States Pharmacopeia (USP) Reference Standard
C3H6O
PHR1219
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
C3H6O
C83201
β-Citronellol, 95%
C10H20O
386529
Chlorotrimethylsilane, purified by redistillation, ≥99%
C3H9ClSi
95541
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
C3H9ClSi
92361
Chlorotrimethylsilane, ≥98.0% (GC)
C3H9ClSi
92360
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
C3H9ClSi
89595
Chlorotrimethylsilane, for GC derivatization, ≥99.0% (GC)
C3H9ClSi
385433
Chlorotrimethylsilane solution, 1.0 M in methylene chloride
C3H9ClSi
384410
Chlorotrimethylsilane solution, 1.0 M in THF
C3H9ClSi
W230901
Citronellol, ≥95%, FCC, FG
C10H20O
W230915
Citronellol, mixture of isomers, natural, ≥95%, FG
C10H20O
15222
N,O-Bis(trimethylsilyl)trifluoroacetamide, for GC derivatization, ≥99.0%
C8H18F3NOSi2
155195
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
C8H18F3NOSi2
T6381
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization
C8H18F3NOSi2
15238
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, contains 1% TMCS, 99% (excluding TMCS)
C8H18F3NOSi2
15209
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, contains 10% TMCS, 98% (excluding TMCS)
C8H18F3NOSi2