EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Corneal biomechanical properties at different corneal cross-linking (CXL) irradiances.
PMID 24677109
Abstract
New corneal cross-linking (CXL) devices are capable of using higher UV-A light irradiances than used in original CXL protocols. The Bunsen-Roscoe law states that a photochemical reaction should stay constant if the delivered total energy is kept constant; however, little clinical data are available to support this hypothesis. We investigated the biomechanical properties of four groups (n = 50 each) of porcine corneas. Three groups were exposed to riboflavin 0.1 % and UV-A irradiation of equal total energy (3 mW/cm(2) for 30 minutes, 9 mW/cm(2) for 10 minutes, and 18 mW/cm(2) for 5 minutes). Controls were exposed to riboflavin 0.1% without irradiation. Young's modulus of 5-mm wide corneal strips was used as an indicator of corneal stiffness. We observed a decreased stiffening effect with increasing UV-A intensity. Young's modulus at 10% strain showed significant differences between 3 mW/cm(2) and 9 mW/cm(2) (P = 0.002), 3 mW/cm(2) and 18 mW/cm(2) (P = 0.0002), 3 mW/cm(2) and the control group (P < 0.0001), and 9 mW/cm(2) and the control group (P = 0.015). There was no difference between 18 mW/cm(2) and the control group (P = 0.064) and between 9 mW/cm(2) and 18 mW/cm(2) (P = 0.503). The biomechanical effect of CXL decreased significantly when using high irradiance/short irradiation time settings. Intrastromal oxygen diffusion capacity and increased oxygen consumption associated with higher irradiances may be a limiting factor leading to reduced treatment efficiency. Our results regarding the efficiency of high-irradiance collagen cross-linking (CXL) raise concerns about the clinical efficiency of the new high-irradiance CXL devices already used in clinical practice without proper validation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V-015
(−)-Riboflavin (Vitamin B2) solution, 100 μg/mL (Methanol:0.1% Ammonium acetate in Water (1:1)), ampule of 1 mL, analytical standard
C17H20N4O6
C9879 Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
C7806 Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
C1188 Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
C8919 Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
C9791 Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
C9301 Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
C0543 Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder (from 0.2 μm-filtered solution), BioReagent, suitable for cell culture
C7521
Collagen from human placenta, Bornstein and Traub Type IV, powder
C4407 Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
C5533
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
C7774 Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
C3657 Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
C8374
Collagen from human placenta, Bornstein and Traub Type IV, powder
H4417 Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, ≥95% (SDS-PAGE)
C5608 Collagen from rabbit skin, Bornstein and Traub Type I, powder
C7661 Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
27666 Collagen from rat tail, for cell biology, soluble
C5483
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
C7624
Collagen human, Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)
C7999
Collagen human, Bornstein and Traub Type I, recombinant, expressed in Nicotiana tabacum (tobacco)
C8993 Collagen ovine, aqueous solution
C6745 Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
C3867 Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
1603006
Riboflavin, United States Pharmacopeia (USP) Reference Standard
C17H20N4O6
Y0000757
Riboflavin for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H20N4O6