EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Emodin-mediated cross-linking enhancement for extracellular matrix homeostasis.
PMID 24680685
Abstract
The extracellular matrix (ECM) is an essential element of mammalian organisms, and its cross-linking formation plays a vital role in ECM development and postnatal homeostasis. Defects in cross-link formation caused by aging, genetic, or environmental factors are known to cause numerous diseases in mammals. To augment the cross-linking formation of ECM, the present study established a ZsGreen reporter system controlled by the promoter of lysyl oxidase-like 1 gene (LOXL1), which serves as both a scaffold element and a cross-linking enzyme in the ECM. By using this system in a drug screen, we identified emodin as a strong enhancer of LOXL1 expression that promoted cross-linking formation of ECM in all the tested systems, including human fibroblast cells, cultured human skin tissues, and animals that received long-term emodin treatment. Collectively, the results suggest that emodin may serve as an effective drug or supplement for ECM homeostasis.